missing translation for 'onlineSavingsMsg'
Learn More

Scopoletin 98.0+%, TCI America™

Catalog No. S03671G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03671G 1 g
S0367100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03671G Supplier TCI America Supplier No. S03671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92-61-5
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00006872
InChI Key RODXRVNMMDRFIK-UHFFFAOYSA-N
Synonym scopoletin, gelseminic acid, 6-methylesculetin, 7-hydroxy-6-methoxy-2h-chromen-2-one, chrysatropic acid, scopoletine, murrayetin, scopoletol, 7-hydroxy-6-methoxycoumarin, 6-o-methylesculetin
PubChem CID 5280460
ChEBI CHEBI:17488
IUPAC Name 7-hydroxy-6-methoxychromen-2-one
SMILES COC1=C(C=C2C(=C1)C=CC(=O)O2)O

Specifications

Melting Point 207°C
Color White-Yellow
Quantity 1 g
Formula Weight 192.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Scopoletin
RTECSNumber GN6930000
TSCA No