missing translation for 'onlineSavingsMsg'
Learn More

Scopoletine 98,0+%, TCI America™

Catalog No. S0367100MG
Change view
Click to view available options
Quantité:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
S0367100MG 100 mg
S03671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0367100MG Supplier TCI America Supplier No. S0367100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 92-61-5
Formule moléculaire C10H8O4
Poids moléculaire (g/mol) 192.17
Numéro MDL MFCD00006872
Clé InChI RODXRVNMMDRFIK-UHFFFAOYSA-N
Synonyme scopoletin, gelseminic acid, 6-methylesculetin, 7-hydroxy-6-methoxy-2h-chromen-2-one, chrysatropic acid, scopoletine, murrayetin, scopoletol, 7-hydroxy-6-methoxycoumarin, 6-o-methylesculetin
PubChem CID 5280460
ChEBI CHEBI:17488
Nom de l’IUPAC 7-hydroxy-6-méthoxychromène-2-1
SOURIRES COC1=C(C=C2C(=C1)C=CC(=O)O2)O

Specifications

Nom ou substance chimique Scopoletin
Point de fusion 207 °C
Couleur Blanc-jaune
Quantité 100 mg
Poids de formule 192.17
Pourcentage de pureté ≥98,0% (HPLC,T)
Forme physique Poudre cristalline
RTECSNumber GN6930000
TSCA Non