missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sclareol

Catalog No. AC465120010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465120010 1 g
AC465120050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465120010 Supplier Thermo Scientific Chemicals Supplier No. 465120010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 515-03-7
Molecular Formula C20H36O2

Specifications

Quantity 1 g
Melting Point 95.0°C to 100.0°C
Boiling Point 218.0°C to 220.0°C (25.0 hPa)
Formula Weight 308.5
InChI Key XVULBTBTFGYVRC-FFADBYAMSA-N
SMILES C[C@@](O)(CC[C@H]1[C@](C)(O)CCC2C(C)(C)CCC[C@]12C)C=C
IUPAC Name (1R,2R,8aS)-1-[(3R)-3-hydroxy-3-methylpent-4-en-1-yl]-2,5,5,8a-tetramethyl-decahydronaphthalen-2-ol
Molecular Weight (g/mol) 308.51
Chemical Name or Material Sclareol
EINECSNumber 208-194-0

RUO – Research Use Only