Learn More
World Precision Spring Scissors, 12 cm, Curved, Extra Fine Tips
Numéro de catalogue. 50822239
Encompass
Spring scissors are a common type of surgical scissor that have a spring action for quick, clean cuts in microsurgery or dissection of delicate tissues. These spring scissors have long (12mm) extra fine blades which gives you greater control in confined spaces. These spring scissors have small, sharp, pointed tips. While they are suitable for use by both left- and right-handed surgeons, they are often preferred by left-handed surgeons. Designed with a symmetrical handle, they are comfortable for ambidextrous use. Left-handed surgeons can use them without any discomfort or awkwardness, unlike some other surgical instruments designed specifically for right-handed use. Left-handed surgeons may find it easier to maneuver the scissors with their left hand, which is typically more dexterous when it comes to fine motor skills. Spring scissors have an ergonomic design for a natural feel with intuitive control.
Numéro de catalogue. 50822239 Fournisseur World Precision Code fournisseur WP14126
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Spring scissors are a common type of surgical scissor that have a spring action for quick, clean cuts in microsurgery or dissection of delicate tissues. These spring scissors have long (12mm) extra fine blades which gives you greater control in confined spaces. These spring scissors have small, sharp, pointed tips. While they are suitable for use by both left- and right-handed surgeons, they are often preferred by left-handed surgeons. Designed with a symmetrical handle, they are comfortable for ambidextrous use. Left-handed surgeons can use them without any discomfort or awkwardness, unlike some other surgical instruments designed specifically for right-handed use. Left-handed surgeons may find it easier to maneuver the scissors with their left hand, which is typically more dexterous when it comes to fine motor skills. Spring scissors have an ergonomic design for a natural feel with intuitive control.