missing translation for 'onlineSavingsMsg'
Learn More

SciMatCo™ Stak-a-Cab™ Stackable Poison Storage Cabinet

Catalog No. 17988475
Encompass
Change view
Click to view available options
Dimensions (D x W x H):
18 x 47 x 18 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H)
17988475 18 x 47 x 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17988475 Supplier SciMatCo™ Supplier No. SC2460

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Make your workspace work for you. SciMat™ Stak-a-CabSciMat™ allows you to design your own storage system and save valuable laboratory space.

  • Constructed of 1in. multi-ply exterior-grade plywood
  • Individual cabinet is designed for stacking or as a stand –alone unit for the laboratory bench
  • Allows for safe compatible organization of your laboratory
  • Poison Cabinet is easily identified by the gray color and a 10 in. Poison label on each door
  • Customize a chemical storage center for your special storage needs
  • Cabinet Floor constructed as a 2in. Liquid tight trough
  • Cabinet supplied with internal metal lock All cabinets constructed to meet or exceed all applicable NFPA and OSHA Standards
  • Floor stand available: Fisher Cat 17-988-487
  • Can be stacked with Stak-A-Cab corrosives, flammables, and combination corrosives/flammables cabinets

Specifications

Load Bearing Capacity 20 x 1 gal.
Certifications/Compliance Meet or exceed all applicable NFPA and OSHA Standards
Dimensions (D x W x H) 18 x 47 x 18 in.
Product Type Poison Cabinet
Material Wood (Plywood)
Color Gray