missing translation for 'onlineSavingsMsg'
Learn More

Schiff's Reagent

Catalog No. AAJ62171AP
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ62171AP 500 mL Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62171AP Supplier Thermo Scientific Chemicals Supplier No. J62171AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 569-61-9 | C19H18ClN3 | 323.824 g/mol

Intended for use in Sigma Periodic Acid Staining Procedure, 395. Kit is intended for use with blood, bone marrow, tissue touch preparations or routine tissue sections. It is used for staining glycoproteins in polyacrylamide gels. Schiff's reagent is also sometimes used to dye organic tissues. It is been widely used for detection of a certain functional group in chemical solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19H18ClN3
Molecular Weight (g/mol) 323.824
MDL Number MFCD00081985
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
Synonym basic red 9, pararosaniline hydrochloride, parafuchsin, pararosaniline chloride, parafuchsine, p-fuchsin, basic fuchsin, ci basic red 9, c.i. basic red 9, parafuksin
PubChem CID 11292
ChEBI CHEBI:87663
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl

Specifications

Packaging Plastic bottle
Quantity 500 mL
Sensitivity Air and light sensitive
Solubility Information It is soluble in water and methanol.
Physical Form Liquid
Chemical Name or Material Schiff's Reagent
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only