missing translation for 'onlineSavingsMsg'
Learn More

Schiff's Reagent, ASTM, Ricca Chemical

Catalog No. R66960001C
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R66960001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R66960001C Supplier Ricca Chemical Company Supplier No. R66960001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Schiff's Reagent
Color Colorless
Name Note ASTM Formulation
CAS 569-61-9, 7631-90-5, 7647-01-0, 7732-18-5
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless
Packaging Amber Glass Bottle
Solvent or Matrix Nonaqueous Solution
pH Acidic
Quantity 1 L
Physical Form Liquid
Show More Show Less