missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCGB1D2 Polyclonal Antibody

Description
SCGB1D2 Polyclonal Antibody for IHC (P), Western Blot
May bind androgens and other steroids, may also bind estramustine, a chemotherapeutic agent used for prostate cancer. May be under transcriptional regulation of steroid hormones.
Spécifications
Spécifications
| Antigène | SCGB1D2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gène | SCGB1D2 |
| Numéro d’acquisition du gène | O95969 |
| Alias du gène | LIPB; LIPHB; Lipophilin-B; LPHB; LPNB; prostatein-like lipophilin B; SCGB1D2; Secretoglobin family 1D member 2; secretoglobin, family 1D member 2; secretoglobin, family 1D, member 2 |
| Symboles de gène | SCGB1D2 |
| Afficher plus de résultats |