missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCAPER Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA566370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA566370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA566370 Fournisseur Invitrogen™ Code fournisseur PA566370
Il en reste null

Rabbit Polyclonal Antibody

Immunogen sequence: KVKAHHTGSTA SSEITPAQSC PPMTVQKASR KNERKDAEGW ETVQRGRPIR SRSTAVMPKV SLATEATRSK DDSDKENVCL LPDESIQKGQ FVGDGTSNTI ESHPKDSLHS CDHPLA Highest antigen sequence identity to the following orthologs - mouse 76%, rat 75%.

CCNA2/CDK2 regulatory protein that transiently maintains CCNA2 in the cytoplasm.

Spécifications

Antigen SCAPER
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCAPER
Gene Accession No. Q9BY12
Gene Alias C430017I08; D530014O03Rik; KIAA1454; KIAA1454-like protein; LOW QUALITY PROTEIN: S phase cyclin A-associated protein in the endoplasmic reticulum; MSTP063; S phase cyclin A-associated protein in the endoplasmic reticulum; S phase cyclin A-associated protein in the ER; SCAPER; S-phase cyclin A associated protein in the ER; S-phase cyclin A-associated protein in the ER; Zfp291; Zinc finger protein 291; znf291
Gene Symbols SCAPER
Host Species Rabbit
Immunogen Recombinant protein fragment of Human SCAPER (Product #RP-107046)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 49855
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats