missing translation for 'onlineSavingsMsg'
Learn More

Scandium Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC ORGSC82Z

Catalog No. ORG-SC8-2Z


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sc1000μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Scandium Oil Standard
7440-20-2 , 64742-65-0
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,99.9
MFCD00016323
Certified Reference Material
[Sc]
scandium
44.96
23952
Oil/Plasma
Triple-leached LDPE bottle
With Certificate of Analysis.
0.1,99.9
Sc
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
SIXSYDAISGFNSX-UHFFFAOYSA-N
Base Oil 75
Petrochemical Testing
50 g
CHEBI:33330

Chemical Identifiers

7440-20-2
44.96
SIXSYDAISGFNSX-UHFFFAOYSA-N
CHEBI:33330
[Sc]
Sc
MFCD00016323
23952
scandium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only