CAS RN 7440-20-2
Scandium foil, 0.127mm (0.005in) thick, 99.9% (REO)
CAS: 7440-20-2 Molecular Formula: Sc Molecular Weight (g/mol): 44.96 MDL Number: MFCD00016323 InChI Key: SIXSYDAISGFNSX-UHFFFAOYSA-N Synonym: unii-yuj4u1ew7r,yuj4u1ew7r,escandio,skandium,atom,powder,ingot,nitrate solution,standard for aas,scandium, lump, vacuum remelted PubChem CID: 23952 ChEBI: CHEBI:33330 IUPAC Name: scandium SMILES: [Sc]
Scandium rod, 12.7mm (0.5in) dia, 99.6% (metals basis excluding Ta)
CAS: 7440-20-2 Molecular Formula: Sc Molecular Weight (g/mol): 44.96 MDL Number: MFCD00016323 InChI Key: SIXSYDAISGFNSX-UHFFFAOYSA-N Synonym: unii-yuj4u1ew7r,yuj4u1ew7r,escandio,skandium,atom,powder,ingot,nitrate solution,standard for aas,scandium, lump, vacuum remelted PubChem CID: 23952 ChEBI: CHEBI:33330 IUPAC Name: scandium SMILES: [Sc]
Scandium arc-cast pellet, 15.9mm (0.63in) dia
CAS: 7440-20-2 Molecular Formula: Sc Molecular Weight (g/mol): 44.96 MDL Number: MFCD00016323 InChI Key: SIXSYDAISGFNSX-UHFFFAOYSA-N Synonym: unii-yuj4u1ew7r,yuj4u1ew7r,escandio,skandium,atom,powder,ingot,nitrate solution,standard for aas,scandium, lump, vacuum remelted PubChem CID: 23952 ChEBI: CHEBI:33330 IUPAC Name: scandium SMILES: [Sc]
Scandium foil, 0.1mm (0.004in) thick, 99.9% (REO)
CAS: 7440-20-2 Molecular Formula: Sc Molecular Weight (g/mol): 44.96 MDL Number: MFCD00016323 InChI Key: SIXSYDAISGFNSX-UHFFFAOYSA-N Synonym: unii-yuj4u1ew7r,yuj4u1ew7r,escandio,skandium,atom,powder,ingot,nitrate solution,standard for aas,scandium, lump, vacuum remelted PubChem CID: 23952 ChEBI: CHEBI:33330 IUPAC Name: scandium SMILES: [Sc]
Scandium ingot, 99.9% (metals basis excluding Ta), Ta nominal 8-12%
CAS: 7440-20-2 Molecular Formula: Sc Molecular Weight (g/mol): 44.96 MDL Number: MFCD00016323 InChI Key: SIXSYDAISGFNSX-UHFFFAOYSA-N Synonym: unii-yuj4u1ew7r,yuj4u1ew7r,escandio,skandium,atom,powder,ingot,nitrate solution,standard for aas,scandium, lump, vacuum remelted PubChem CID: 23952 ChEBI: CHEBI:33330 IUPAC Name: scandium SMILES: [Sc]
Scandium pieces, distilled dendritic, 99.9% (REO)
CAS: 7440-20-2 Molecular Formula: Sc Molecular Weight (g/mol): 44.96 MDL Number: MFCD00016323 InChI Key: SIXSYDAISGFNSX-UHFFFAOYSA-N Synonym: unii-yuj4u1ew7r,yuj4u1ew7r,escandio,skandium,atom,powder,ingot,nitrate solution,standard for aas,scandium, lump, vacuum remelted PubChem CID: 23952 ChEBI: CHEBI:33330 IUPAC Name: scandium SMILES: [Sc]
Scandium, plasma standard solution, Specpure™ Sc 1000μg/mL
CAS: 7440-20-2 Molecular Formula: Sc Molecular Weight (g/mol): 44.96 MDL Number: MFCD00016323 InChI Key: SIXSYDAISGFNSX-UHFFFAOYSA-N PubChem CID: 23952 ChEBI: CHEBI:33330 IUPAC Name: scandium SMILES: [Sc]
Scandium foil, 0.025mm (0.001in) thick, 99% (REO)
CAS: 7440-20-2 Molecular Formula: Sc Molecular Weight (g/mol): 44.96 MDL Number: MFCD00016323 InChI Key: SIXSYDAISGFNSX-UHFFFAOYSA-N Synonym: unii-yuj4u1ew7r,yuj4u1ew7r,escandio,skandium,atom,powder,ingot,nitrate solution,standard for aas,scandium, lump, vacuum remelted PubChem CID: 23952 ChEBI: CHEBI:33330 IUPAC Name: scandium SMILES: [Sc]
Scandium foil, 0.25mm (0.01in) thick, 99.9% (REO)
CAS: 7440-20-2 Molecular Formula: Sc Molecular Weight (g/mol): 44.96 MDL Number: MFCD00016323 InChI Key: SIXSYDAISGFNSX-UHFFFAOYSA-N Synonym: unii-yuj4u1ew7r,yuj4u1ew7r,escandio,skandium,atom,powder,ingot,nitrate solution,standard for aas,scandium, lump, vacuum remelted PubChem CID: 23952 ChEBI: CHEBI:33330 IUPAC Name: scandium SMILES: [Sc]
Scandium Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™
CAS: 7440-20-2 Molecular Formula: Sc Molecular Weight (g/mol): 44.96 MDL Number: MFCD00016323 InChI Key: SIXSYDAISGFNSX-UHFFFAOYSA-N PubChem CID: 23952 ChEBI: CHEBI:33330 IUPAC Name: scandium SMILES: [Sc]
Scandium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material