missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SB 699551
Description
Selective 5-ht5a receptor antagonist (pKi values are 8.3, < 6.0, < 6.0, < 6.0, < 5.5 and < 5.5 for 5-ht5a, 5-HT1B/D, 5-HT2A, 5-HT2C, 5-HT1A and 5-HT7 receptors respectively).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 584.66 |
| Purity | >99% |
| Molecular Weight (g/mol) | 584.67 |
| Chemical Name or Material | SB 699551 |
Safety and Handling
| Recommended Storage | Store at +4°C |