accessibility menu, dialog, popup

UserName

CH3NH3CNNHOHClHCl

CAS RN 864741-95-7

IUPAC Name: 3-cyclopentyl-N-[2-(dimethylamino)ethyl]-N-[(4'-{[(2-phenylethyl)amino]methyl}-[1,1'-biphenyl]-4-yl)methyl]propanamide dihydrochloride
Synonyms: 3-cyclopentyl-N-[2-(dimethylamino)ethyl]-N-[(4'-{[(2-phenylethyl)amino]methyl}-[1,1'-biphenyl]-4-yl)methyl]propanamide dihydrochloride
Molecular Weight (g/mol): 584.67
Molecular Formula: C34H47Cl2N3O
InChi Key: QJMKBIHLMPTYTI-UHFFFAOYSA-N
SMILES: Cl.Cl.CN(C)CCN(CC1=CC=C(C=C1)C1=CC=C(CNCCC2=CC=CC=C2)C=C1)C(=O)CCC1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results