missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 505124

Catalog No. 326310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
326310 10 mg
326350 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 326310 Supplier Tocris Bioscience™ Supplier No. 3263/10
Only null left

Selective inhibitor of TGF-βRI, ALK4 and ALK7

Selective inhibitor of transforming growth factor-β type I receptor (ALK5), ALK4 and ALK7 (IC50 values are 47 and 129 nM for ALK5 and ALK4 respectively). Selectively inhibits signaling from TGF-β and activin; does not inhibit other ALK family members.

Chemical Identifiers

CAS 694433-59-5
Molecular Formula C20H21N3O2

Specifications

Quantity 10 mg
Formula Weight 335.4
Purity >98%
Molecular Weight (g/mol) 335.407
Chemical Name or Material SB 505124
Recommended Storage Store at -20°C