accessibility menu, dialog, popup

UserName

CH3NNNHCH3CH3CH3OO

CAS RN 694433-59-5

IUPAC Name: 2-[4-(2H-1,3-benzodioxol-5-yl)-2-tert-butyl-1H-imidazol-5-yl]-6-methylpyridine
Synonyms: 2-[5-(2H-1,3-benzodioxol-5-yl)-2-tert-butyl-3H-imidazol-4-yl]-6-methylpyridine
Molecular Weight (g/mol): 335.41
Molecular Formula: C20H21N3O2
InChi Key: WGZOTBUYUFBEPZ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=N1)C1=C(N=C(N1)C(C)(C)C)C1=CC2=C(OCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results