Learn More
Abcam Inc SB 202190, cell-permeable p38 MAPK inhibitor, 50MG
Numéro de catalogue. 502707404
Encompass
MW 331.3 Da, Purity >99%. Highly selective, potent and cell-permeable p38 MAPK inhibitor (IC values are 50 and 100 nM at SAPK2a/p38 and SAPK2b/p382, respectively). Selective over a wide range of other kinases. Apoptosis and autophagy inducer. 3D Growth matrix component and additive for long term growth; also component of gastric organoid culture media.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502707404 Fournisseur Abcam Inc Code fournisseur AB12063850MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 331.3 Da, Purity >99%. Highly selective, potent and cell-permeable p38 MAPK inhibitor (IC values are 50 and 100 nM at SAPK2a/p38 and SAPK2b/p382, respectively). Selective over a wide range of other kinases. Apoptosis and autophagy inducer. 3D Growth matrix component and additive for long term growth; also component of gastric organoid culture media.
The product is subject to the following: Abcam Restricted Use Statement