missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SB 202190, 99+%
Inhibits p38 kinase activity in vivo through competition with ATP | CAS: 152121-30-7 | C20H14FN3O, C20H14FN3O | 331.35 g/mol
$270.23 - $527.95
Chemical Identifiers
| CAS | 152121-30-7 |
|---|---|
| Molecular Formula | C20H14FN3O |
| Synonym | 4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)imidazole |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ60950MB
|
N/A | 25 mg |
Each for $270.23
|
|
|||||
|
AAJ60950MF
|
N/A | 50 mg |
Each for $527.95
|
|
|||||
Description
Inhibits p38 kinase activity through competition with ATP
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 152121-30-7 | |
| C20H14FN3O | |
| 4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)imidazole |
Specifications
| Yellow | |
| Inhibits p38 kinase activity in vivo through competition with ATP | |
| C20H14FN3O | |
| 4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)imidazole | |
| Soluble in DMSO at 30mg/ml | |
| Crystalline solid | |
| FC1=CC=C(C=C1)C1=C(NC(N1)=C1C=CC(=O)C=C1)C1=CC=NC=C1 | |
| 331.35 |
| -30°C to -10°C | |
| 152121-30-7 | |
| 331.35g/mol | |
| 240°C to 243°C | |
| Signal Transduction Reagent | |
| NJNKPVPFGLGHPA-UHFFFAOYSA-N | |
| 4-[4-(4-fluorophenyl)-5-(pyridin-4-yl)-2,3-dihydro-1H-imidazol-2-ylidene]cyclohexa-2,5-dien-1-one | |
| ≥99% |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only