missing translation for 'onlineSavingsMsg'
Learn More

SB 202190, 99+%

Catalog No. AAJ60950MB
Change view
Click to view available options
Quantity:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60950MB 25 mg
AAJ60950MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60950MB Supplier Thermo Scientific Chemicals Supplier No. J60950MB

Inhibits p38 kinase activity in vivo through competition with ATP | CAS: 152121-30-7 | C20H14FN3O, C20H14FN3O | 331.35 g/mol

Inhibits p38 kinase activity through competition with ATP

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 152121-30-7
Molecular Formula C20H14FN3O
Synonym 4-(4-Fluorophenyl)-2-(4-hydroxyphenyl)-5-(4-pyridyl)imidazole

Specifications

Quantity 25 mg
Description Yellow
Content And Storage -30°C to -10°C
For Use With (Application) Inhibits p38 kinase activity in vivo through competition with ATP
Formula Weight 331.35g/mol
Melting Point 240°C to 243°C
Solubility Soluble in DMSO at 30mg/ml
Product Type Signal Transduction Reagent
Form Crystalline solid
InChI Key NJNKPVPFGLGHPA-UHFFFAOYSA-N
SMILES FC1=CC=C(C=C1)C1=C(NC(N1)=C1C=CC(=O)C=C1)C1=CC=NC=C1
IUPAC Name 4-[4-(4-fluorophenyl)-5-(pyridin-4-yl)-2,3-dihydro-1H-imidazol-2-ylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol) 331.35
Assay ≥99%
Show More Show Less
Recommended Storage Store at -20°C

RUO – Research Use Only