missing translation for 'onlineSavingsMsg'
Learn More

Savory Oil, Summer Variety, Spectrum™ Chemical

Catalog No. 18613082
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613082 500 mL Amber Glass Bottle
18613080 25 mL Amber Glass Bottle
18613081 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18613082 Supplier Spectrum Chemical Mfg Cor Supplier No. S1096500ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8016-68-0

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Grade Reagent
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Savory Oil, Summer Variety

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.