missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant™ SpeedVac™ Medium Capacity Vacuum Concentrators for Combinatorial Chemistry Applications

Catalog No. 13875374
Change view
Click to view available options
Description:
SpeedVac SPD140 Vacuum Concentrator Kit
SpeedVac SPD140DDA Vacuum Concentrator
Electrical Requirements:
115 V 60 Hz
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Electrical Requirements
13875374 SpeedVac SPD140 Vacuum Concentrator Kit 115 V 60 Hz
13875345 SpeedVac SPD140DDA Vacuum Concentrator 115 V 60 Hz
13875372 SpeedVac SPD140 Vacuum Concentrator Kit 115 V 60 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13875374 Supplier Thermo Scientific™ Supplier No. SPD140P2115

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.

Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.

SpeedVac SPD140DDA is a medium capacity vacuum concentrator used with a wide variety of solvents and offer resistance to TFA, DSMO and other aggressive solvents used in combinatorial chemistry applications.   It is available in kit for ease of ordering.

SPD140DDA Features:

  • All the product features of SPD130DLX PLUS:
  • Radiant lamps within the chamber
  • Heated lid for processing high boiling points (DMSO,DMF)
  • Independent timers for both heat and run
  • Audible alarm to alert user when operation is finished
  • Built-in vacuum gauge and vacuum display
  • Wider vapor port and tubing for enhanced vapor throughput - reducing drying times
  • Adjustable vacuum level for preventing bumping of difficult solvent mixtures and optimizing drying times
  • -5°C Cold Trap specially designed for trapping high boiling point solvents

Solvent and Solvent Combinations for SPD140P1 KIT

  • Acetic acid
  • Ammonium hydroxide
  • Formic acid
  • Hydrochloric acid
  • TFA
  • Methylene chloride and TFA
  • Acetone and HCL
  • Methylene chloride and MEOH
  • DMSO and methanol

Solvent Combinations for SPD140 P2 KIT

  • DMSO
  • DMF
  • Toluene

SPD140  P1 includes:

  • Acid-Resistant SpeedVac™ Vacuum Concentrator
  • -104°C Ultra-Low Temperature Refrigerated Vapor Trap
  • Oil-Free ETFE/PFA 4-Headed Pump
  • 40 x 1.5-2.0ml Glass Tubes Rotor (RH40-11)
  • Glass Condensation Flask
  • Tubing and Fitting Kit
  • CryoCool™ Heat Transfer Fluid (1L)

SPD140P2 includes:

  • Acid-Resistant SpeedVac™ Vacuum Concentrator
  • -50°C Universal Vacuum Source
  • 24 x 1 dram vials (15 x 45 mm) 4ml - dram vial Rotor (RH24-15)
  • Chemical Trap
  • Tubing and Fitting Kit
  • Disposable Cartridge
  • Glas Condensation Flask
  • Vacuum Pump Lubricating Oil
  • CryoCool™ Heat Transfer Fluid (1L)

SPD140 P1 and P2 kits are recommended for:

  • Solid phase extractions
  • Drying lipid extractions
  • Flash chromatography fractions
  • Solid phase synthesis cleavage solutions such as protein hydrolyzates and evaporating synthesis solids
  • Compound management
  • Evaporating dissolution solventsRotors are available for a wide variety of applications and sample formats. Please check the rotor list.

Typical applications SPD140DDA:

Requires:

  • Drying lipid extraction
  • Flash chromatography fractions
  • Combinatorial Chemistry
  • Solid phase synthesis cleavage solutions such as protein hydrolyzates and evaporating synthesis solids
  • Removing ethyl acetate from samples for CAT assay
  • Drug metabolites in solid phase extracts
  • Analyte elution solvents for evaporation

Specifications

Certifications UL Listed to US and Canadian requirements. CE marked. Please contact Customer Service for possible alternatives.
Description SpeedVac SPD140 Vacuum Concentrator Kit
Electrical Requirements 115 V 60 Hz