missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ Turbo 15 PES Centrifugal Concentrators

Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.

$247.44 - $948.54

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
Sample Volume 15 mL
View More Specs

Products 12
Catalog Number Mfr. No. MWCO Quantity Price Quantity & Availability  
Catalog Number Mfr. No. MWCO Quantity Price Quantity & Availability  
14558658
View Documents Promotion Details
Sartorius
VS15T91
3 kDa 12/Pk.
Pack of 12 Each for $247.44
Only null left
 
14558655
View Documents Promotion Details
Sartorius
VS15T41
100 kDa 12/Pk.
Pack of 12 Each for $247.44
Only null left
 
14558652
View Documents Promotion Details
Sartorius
VS15T11
5 kDa 12/Pk.
Pack of 12 Each for $247.44
Only null left
 
14558654
View Documents Promotion Details
Sartorius
VS15T31
50 kDa 12/Pk.
Pack of 12 Each for $247.44
Only null left
 
14558651
View Documents Promotion Details
Sartorius
VS15T01
10 kDa 12/Pk.
Pack of 12 Each for $247.44
Only null left
 
14558653
View Documents Promotion Details
Sartorius
VS15T21
30 kDa 12/Pk.
Pack of 12 Each for $247.44
Only null left
 
14558660
View Documents Promotion Details
Sartorius
VS15T02
10 kDa 48/Pk.
Pack of 48 Each for $948.54
Only null left
 
14558661
View Documents Promotion Details
Sartorius
VS15T12
5 kDa 48/Pk.
Pack of 48 Each for $948.54
Only null left
 
14558662
View Documents Promotion Details
Sartorius
VS15T22
30 kDa 48/Pk.
Pack of 48 Each for $948.54
Only null left
 
14558664
View Documents Promotion Details
Sartorius
VS15T42
100 kDa 48/Pk.
Pack of 48 Each for $948.54
Only null left
 
14558663
View Documents Promotion Details
Sartorius
VS15T32
50 kDa 48/Pk.
Pack of 48 Each for $948.54
Only null left
 
14558667
View Documents Promotion Details
Sartorius
VS15T92
3 kDa 48/Pk.
Pack of 48 Each for $948.54
Only null left
 
Description

Description

Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.

Twin vertical membranes and a sleek internal design for ultra-fast concentration
  • In a single spin, 15 ml solutions can be concentrated up to 150x in 10-30 minutes
  • Large, high-flux PES membranes feature ultra-low protein binding and low extractables
  • High, constant filtration pressure allows optimal crossflow to minimize membrane fouling
Designed for maximum convenience, safety and sample recovery
  • Angular, pipette-friendly pocket facilitates easy retentate recovery of greater than 95%
  • Eliminates the risk of sample filtration to dryness
  • Stable, inert polystyrene housing with high chemical compatibility (pH 1-14)
  • UV joining technology for a smooth transition between membrane and housing
  • Volume graduation for easy monitoring of concentrate volume and 100μl dead-stop volume
Universal compatibility for rotors that accept 50mL conical bottom tubes
  • Handles sample volumes up to 15mL in swing bucket rotors (11mL in fixed angle rotors)
  • Designed for rotors that accommodate Falcon 50mL conical tubes:
    • Beckman Allegra 25R with TS-5.1-500 swing-out rotor, BUC 5 buckets, adaptors
    • Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors
    • Multifuge 3 S-R with 75006445 rotor, 75006441 buckets, adaptors
Specifications

Specifications

Single-use
Centrifuge
15 mL
7.2 cm2
Polystyrene
Centrifugal Concentrator
11.8 cm
Ultrafiltration
PES
Non-sterile
Concentration, desalting or buffer exchange applications
4,000 x g
11 mL
27 mm
Videos
SDS
Documents

Documents