missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ Turbo 15 PES Centrifugal Concentrators

Catalog No. 14558658
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
5 kDa
50 kDa
Quantity:
12/Pk.
48/Pk.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity
14558658 3 kDa 12/Pk.
14558655 100 kDa 12/Pk.
14558652 5 kDa 12/Pk.
14558654 50 kDa 12/Pk.
14558651 10 kDa 12/Pk.
14558653 30 kDa 12/Pk.
14558660 10 kDa 48/Pk.
14558661 5 kDa 48/Pk.
14558662 30 kDa 48/Pk.
14558664 100 kDa 48/Pk.
14558663 50 kDa 48/Pk.
14558667 3 kDa 48/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 14558658 Supplier Sartorius Supplier No. VS15T91
Only null left

Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.

Twin vertical membranes and a sleek internal design for ultra-fast concentration
  • In a single spin, 15 ml solutions can be concentrated up to 150x in 10-30 minutes
  • Large, high-flux PES membranes feature ultra-low protein binding and low extractables
  • High, constant filtration pressure allows optimal crossflow to minimize membrane fouling
Designed for maximum convenience, safety and sample recovery
  • Angular, pipette-friendly pocket facilitates easy retentate recovery of greater than 95%
  • Eliminates the risk of sample filtration to dryness
  • Stable, inert polystyrene housing with high chemical compatibility (pH 1-14)
  • UV joining technology for a smooth transition between membrane and housing
  • Volume graduation for easy monitoring of concentrate volume and 100μl dead-stop volume
Universal compatibility for rotors that accept 50mL conical bottom tubes
  • Handles sample volumes up to 15mL in swing bucket rotors (11mL in fixed angle rotors)
  • Designed for rotors that accommodate Falcon 50mL conical tubes:
    • Beckman Allegra 25R with TS-5.1-500 swing-out rotor, BUC 5 buckets, adaptors
    • Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors
    • Multifuge 3 S-R with 75006445 rotor, 75006441 buckets, adaptors

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
MWCO 3 kDa
Sample Volume 15 mL
Sterility Non-sterile
Filtration Area 7.2 cm2
For Use With (Application) Concentration, desalting or buffer exchange applications
Housing Material Polystyrene
Max. Force 4,000 x g
Quantity 12/Pk.
Type Centrifugal Concentrator
Volume (Metric) 11 mL
Length (Metric) 11.8 cm
Width (Metric) 27 mm
Show More Show Less