missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Vivaspin™ 6 Centrifugal Concentrators
Description
Developed to offer increased volume flexibility and performance
- Compact tubes can process up to an impressive 6mL initial volume
- Optimally suited for the concentration of antibody, protein, nanoparticle and virus samples
- Universal rotor compatibility:
- Use in either swing bucket or fixed angle rotors accepting standard 15mL conical bottom test tubes
- Features premium, high-throughput PES membranes
- Twin vertical membrane design lead to unparalleled filtration speed and over 100-fold concentration
- The dead stop pocket prevents concentration to dryness for safe sample concentration
- Nearly complete concentrate recovery of over 90%
- Remaining volume is easy to see with the printed graduations and clear housing
- Modified dead stop pocket further simplifies direct pipette recovery of the final concentrate
Spécifications
Spécifications
| Disposable | Single-use |
| Filter Type | Ultrafiltration |
| For Use With (Equipment) | Centrifuge |
| Material (Membrane) | PES |
| MWCO | 3 kDa |
| Sample Volume | 6 mL |
| Sterility | Non-sterile |
| Filtration Area | 2.58 cm2 |
| Housing Material | Polycarbonate |
| Quantity | 100/Pk. |
| Afficher plus de résultats |