Learn More
Sartorius Glass Filtration Holder
This glass vacuum filtration holder has been designed for particle testing, clarification and sterile filtration with the use of 24 or 25 mm filters.
Supplier:  Sartorius 16306
 View more versions of this product
Catalog No. 14-555-544
Only null left
Description
Description
  • Vaccuum Filter Holder is delivered with a glass frit filter support that ensures a uniform distribution of the retained particles; therefore it is recommended when the residue on the filter surface is of interest.
  • Millex Filter is delivered with a PTFE-coated screen filter support that has the benefit to be easy to clean. Therefore it is recommended when the filtrate is required or when liquids that are difficult to remove from glass supports must be examined.
Specifications
Specifications
Reusable
Glass
3 cm2
Vacuum Filter Holder
2.5 cm
Vacuum Filter Holder, Manifold System
Sterile
Each
30 mL
Product Suggestions
Product Suggestions