Learn More
Acrobiosystems ACROBiosystems SARS-CoV-2SpikeRBD TiterAssayKit DFS Item
Supplier:  ACROBiosystems Inc RASA02196TESTS
Kits;96TESTS;This assay kit employs a standard sandwich-ELISA format, providing a rapid detection of SARS-CoV-2 Spike RBD. The kit consists of microplate pre-coated with Anti-SARS-CoV-2 Spike RBD Antibody, an SARS-CoV-2 Spike RBD as Control, an biotin-Anti-SARS-CoV-2 Spike RBD Antibody, HRP-Streptavidin and buffers.;This assay kit employs a standard sandwich-ELISA format providing a rapid detection of SARS-CoV-2 Spike RBD. The kit consists of pre-coated Anti-SARS-CoV-2 Spike RBD Antibody Microplate, SARS-CoV-2 Spike RBD as Control, biotin-Anti-SARS-CoV-2 Spike RBD antibody and HRP-Streptavidin. Detection is performed using HRP-Streptavidin with sensitivity of 12 pg/mL (QC tested).
Catalog No. 50-210-7432
Only null left
Explore available promotions