missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS Coronavirus Spike Protein Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA141165

Numéro de catalogue. 01675739


Il en reste null

Description

Description

This antibody has been used for Flow Cytometry to detect HEK293T.17 cells transfected with vaccine-expressing membrane-bound SARS-CoV-2 spike protein. View Article The immunogen is 100% (16/16) homologous to SARS-COV-2 (COVID-19) Spike protein. The cross reactivity is not empirically tested. Suggested positive control: transfected mouse melanoma cell lysate. Immunogen sequence: GNCDVVIGIV NNTVYDP.

It has recently been shown that SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The glycosylated spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients.
Spécifications

Identifiants chimiques

SARS Coronavirus Spike Protein
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Virus
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Primary
Antigen affinity chromatography
1.0 mg/mL
Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.02% sodium azide
Synthetic peptide corresponding to amino acids 1124-1140 from the S (Spike glycoprotein) (Spike protein S2') for the Human SARS coronavirus.
Antibody

Spécifications

SARS Coronavirus Spike Protein
Polyclonal
Unconjugated
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Synthetic peptide corresponding to amino acids 1124-1140 from the S (Spike glycoprotein) (Spike protein S2') for the Human SARS coronavirus.
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1.0 mg/mL
PBS with 0.02% sodium azide
Rabbit
Antigen affinity chromatography
RUO
Virus
Antibody
IgG
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions