Learn More
Polysciences Inc SAR-GEL Water Indicating Paste, 12 x 1 oz tubes
Numéro de catalogue. NC0191344
Encompass
Water indicating paste provides a fast, reliable way to detect water bottoms in storage tanks containing gasoline and gasoline/alcohol blends, diesel, jet fuel, fuel oil, solvents and other materials.
Numéro de catalogue. NC0191344 Fournisseur Polysciences Inc Code fournisseur 2461512
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Water indicating paste provides a fast, reliable way to detect water bottoms in storage tanks containing gasoline and gasoline/alcohol blends, diesel, jet fuel, fuel oil, solvents and other materials.