missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Saquinavir mesylate
Description
Saquinavir mesylate is an inhibitor of human immunodeficiency virus (HIV) protease (Ki values are <0.1 and 0.12 nM for HIV-2 and HIV-1 protease respectively). Exhibits high antiviral activity and low cytotoxicity. In silico docking models predict potential as an inhibitor of SARS-CoV-2 3CLpro.
Spécifications
Spécifications
| Quantity | 10 mg |
| Formula Weight | 766.95 |
| Purity | >99% |
| Molecular Weight (g/mol) | 766.955 |
| Chemical Name or Material | Saquinavir mesylate |
Sécurité et manipulation
| Recommended Storage | Store at +4°C |