missing translation for 'onlineSavingsMsg'
Learn More

Sand, MilliporeSigma™

Catalog No. 50947448
Change view
Click to view available options
Quantity:
22.72 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50947448 22.72 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947448 Supplier Technidata Supplier No. SX007630

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Plastic Drum
CAS 14808-60-7
pH 5 to 8 (400 g/L, H2O, 20°C)
Melting Point 1713°C
Boiling Point >2200°C (1013 hPa)
Molecular Weight (g/mol) 60.08
Quantity 22.72 kg