missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120mL (4 oz.) 53mm Specimen Containers
GREENER_CHOICE

Catalog No. 1371156
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Closure Color:
Orange
White
Yellow
Label:
No Label
Non-tabbed
Tabbed
No. per Pack:
300/Pk., Bagged Separately
75/Pk., Pre-assembled
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Closure Color Label No. per Pack
1371156 Sterile Orange Tabbed 75/Pk., Pre-assembled
1371157 Non-sterile Yellow Non-tabbed 75/Pk., Pre-assembled
22610141 Non-sterile White Non-tabbed 75/Pk., Pre-assembled
22610146 Non-sterile Orange No Label 300/Pk., Bagged Separately
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 1371156 Supplier Thermo Scientific™ Supplier No. 01 0001
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prevent bio-hazardous spills with leak-resistant wide-mouth containers, ideally suited for urine collection from women during pregnancy and for older or overweight patients.

Prevent bio-hazardous spills with the proprietary, all plastic, leak-resistant seal of Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 120 mL (4 oz.) 53mm Specimen Containers. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.

Features:

  • 120mL (4 oz.) 53mm
  • Bio-Tite caps create leak-resistant seal for superior safety and handling
  • Dual-threaded caps create improved sealing
  • Sterilized by gamma irradiation, which eliminates potential contamination
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Stackable containers are easy to store, saving lab space
  • Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
  • Sterile indicator tab assures tamper evidence
  • Bilingual English and Spanish label facilitates accurate usage

Ordering Information:
Case of 300

Specifications

Product Type Specimen Container
Closure Included Yes
Material Virgin Polypropylene
Shape Cylindrical
Sterility Sterile
Capacity (Metric) 120 mL
Closure Color Orange
Closure Size 53 mm
Closure Type Bio-Tite Lid
Description Cap and container pre-assembled
Diameter (Metric) 53 mm
Label Tabbed
No. per Case 300/Cs.
No. per Pack 75/Pk., Pre-assembled
Show More Show Less