missing translation for 'onlineSavingsMsg'
Learn More

Samarium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)

Catalog No. AA40297HF
Change view
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40297HF 10 mm
AA40297DH 25 mm
AA40297DJ 50 mm
AA40297DM 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA40297HF Supplier Thermo Scientific Chemicals Supplier No. 040297HF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-19-9 | Sm | 150.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]

Specifications

Physical Form Cast Rod
Quantity 10 mm
Odor Odorless
Packaging Mylar under Argon
Percent Purity 99.9%
Assay (metals basis excluding Ta)
Chemical Name or Material Samarium rod, 6.35mm (0.25 in.) dia.
Hazard Category H228-H260
Precautionary Statement P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: III
EINECSNumber 231-128-7
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only