missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Samarium AA Standard, 1000 ppm Sm in 3% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 93.0, 4.5, 0.11 |
| CAS Max % | 97.0, 4.68, 0.12 |
| Odor | Odorless |
| Concentration or Composition (by Analyte or Components) | 1mL = 1mg Sm (1, 000ppm Sm)Sm2O3 in 3% HNO3 |
| Chemical Name or Material | Samarium AA Standard |
Safety and Handling
| ShelfLife | 24 months |