missing translation for 'onlineSavingsMsg'
Learn More

Samarium, 99.9%, (trace metal basis), 20 to 40 mesh

Catalog No. AC294780500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294780500 50 g Plastic bottle
AC294780100 10 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC294780500 Supplier Thermo Scientific Chemicals Supplier No. 294780500

CAS: 7440-19-9 | Sm | 150.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]

Specifications

Boiling Point 1778.0°C
Melting Point 1072.0°C
Color Gray
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.9%
Linear Formula Sm
UN Number 3132
Fieser 17,305
Merck Index 15, 8484
Solubility Information Solubility in water: reacts. Other solubilities: forming hydrogen
Formula Weight 150.36
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Samarium, 40 Mesh
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
In contact with water releases flammable gas.
Flammable solid.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a dry place.
Store in a closed container.
In case of fire: Use dry sand,dry chemical or alcohol-resistant foam for extinction.
EINECSNumber 231-128-7

RUO – Research Use Only