missing translation for 'onlineSavingsMsg'
Learn More

Salicylic acid, 99+%, ACS reagent

Catalog No. AC419225000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419225000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC419225000 Supplier Thermo Scientific Chemicals Supplier No. 419225000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69-72-7 | C7H6O3 | 138.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 69-72-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002439
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
Synonym salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
PubChem CID 338
ChEBI CHEBI:16914
IUPAC Name 2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC=C1O

Specifications

Melting Point 158.0°C to 161.0°C
Boiling Point 211.0°C (20.0 mmHg)
Color White
Quantity 500 g
Flash Point 157°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Powder
Assay Percent Range 99+%
Residue after Ignition 0.01% max.
Vapor Pressure 0.3mbar at 95°C
Beilstein 10, 43
Merck Index 15, 8469
Formula Weight 138.12
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Salicylic acid
Show More Show Less
Hazard Category SIGNAL WORD: DANGER!
Hazard Statement GHS P Statement Harmful if swallowed. Causes serious eye damage.
Precautionary Statement Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health: 2
Flammability: 1
Instability: 0

RUO – Research Use Only