missing translation for 'onlineSavingsMsg'
Learn More

Salicylic Acid, Crystal, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18606151
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606151 500 g Amber Glass Bottle
18606152 2.5 kg Poly Pail
18606153 12 kg Fiber Drum
18606154 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606151 Supplier Spectrum Chemical Mfg Cor Supplier No. SA125500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

SA125, 69-72-7, C7H6O3

Spectrum™ Chemical Salicylic Acid, Crystal, USP is a topical application used to treat skin conditions that involve scaling or overgrowth of skin cells such as psoriasis, dandruff, corns, calluses, and warts. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 69-72-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002439
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
IUPAC Name 2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC=C1O

Specifications

CAS Max % 1
Percent Purity 98 to 102%
Grade USP
Quantity 500 g
Ignition Residue 0.0005
Loss on Drying 0.005
Packaging Amber Glass Bottle