Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salicylic Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Specifications
Specifications
| Quantity | 500 g |
| Flash Point | 157.00°C |
| Packaging | Wide Mouth Amber Glass Bottle |
| Physical Form | Crystalline |
| Specific Gravity | 1L = 1.44kg |
| Formula Weight | 138.12 |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Chemical Name or Material | Salicylic Acid |