missing translation for 'onlineSavingsMsg'
Learn More

Salbutamol Hemisulfate 98.0+%, TCI America™

Catalog No. S05315G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05315G 5 g
S05311G 1 g
S053125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S05315G Supplier TCI America Supplier No. S05315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51022-70-9
Molecular Formula C13H23NO7S
Molecular Weight (g/mol) 337.387
MDL Number MFCD00055200
InChI Key OVICLFZZVQVVFT-UHFFFAOYSA-N
Synonym salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate
PubChem CID 9884233
IUPAC Name 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid
SMILES CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 5 g
Formula Weight 288.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Salbutamol Hemisulfate
RTECSNumber CZ6430100
TSCA No