missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SAHA

Catalog No. 465210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
465210 10 mg
465250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 465210 Supplier Tocris Bioscience™ Supplier No. 4652/10
Only null left

Class I and II HDAC inhibitor

SAHA inhibits Class I and II histone deacetylases (HDACs); induces accumulation of acetylated histones H2A, H2B, H3 and H4 in transformed cultured cells. Suppresses cell growth in a range of cancer cell lines; induces apoptosis in cutaneous T cell lymphoma cells in vitro. Activates autophagy. SAHA increases efficiency of transcription factor-induced reprogramming of mouse embryonic fibroblasts (MEF) to induced pluripotent stem cells (iPSC). Also enhances adeno-associated virus transduction of HeLa cells.

Chemical Identifiers

CAS 149647-78-9
Molecular Formula C14H20N2O3

Specifications

Quantity 10 mg
Formula Weight 264.32
Purity >98%
Molecular Weight (g/mol) 264.325
Chemical Name or Material SAHA
Recommended Storage Store at -20°C