CAS RN 149647-78-9
IUPAC Name:
N-hydroxy-N'-phenyloctanediamide
Synonyms:
vorinostat vorinostat (product) M344 compound nhnpoda vorinostat (substance) SHH zolinza suberanilohydroxamic acid SAHA saha cpd
Molecular Weight (g/mol):
264.33
Molecular Formula:
C14H20N2O3
InChi Key:
WAEXFXRVDQXREF-UHFFFAOYSA-N
SMILES:
ONC(=O)CCCCCCC(=O)NC1=CC=CC=C1
1
–
2
of
2
results
1
N-Hydroxy-N'-phenyloctanediamide 98.0+%, TCI America™
CAS: 149647-78-9 Molecular Formula: C14H20N2O3 Molecular Weight (g/mol): 264.325 MDL Number: MFCD00945317 InChI Key: WAEXFXRVDQXREF-UHFFFAOYSA-N Synonym: vorinostat,saha,suberoylanilide hydroxamic acid,zolinza,n-hydroxy-n'-phenyloctanediamide,suberanilohydroxamic acid,n1-hydroxy-n8-phenyloctanediamide,saha cpd,vorinostat usan,octanediamide, n-hydroxy-n'-phenyl PubChem CID: 5311 ChEBI: CHEBI:45716 IUPAC Name: N'-hydroxy-N-phenyloctanediamide SMILES: C1=CC=C(C=C1)NC(=O)CCCCCCC(=O)NO
2
R&D Systems™ SAHA
CAS: 149647-78-9 Molecular Formula: C14H20N2O3