missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Safranine O

Catalog No. AAB2167418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2167418 50 g
AAB2167409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2167418 Supplier Thermo Scientific Chemicals Supplier No. B2167418
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 477-73-6 | C20H19ClN4 | 350.85 g/mol

It is a redox and adsorption indicator. Employed as a biological stain used in histology, cytology, bacteriology. It is commonly used as a counter stain for gram-negative bacteria in the Gram stain procedure. It has been observed to bind to DNA at a pH 2.21. In the Flemming triple stain procedure it is used to stain for chromosomes. In late Paleozoic pteridosperm cuticles it has shown increased contrast, emphasized cutinization differences, and enhanced the three-dimensional morphology of complex epidermal features when compared with other dyes. The compound has also been used with alcian blue in the histochemical study of the distribution and morphometric characteristics of mast cells. It has also been used to detect mucin and cartilage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a redox and adsorption indicator. Employed as a biological stain used in histology, cytology, bacteriology. It is commonly used as a counter stain for gram-negative bacteria in the Gram stain procedure. It has been observed to bind to DNA at a pH 2.21. In the Flemming triple stain procedure it is used to stain for chromosomes. In late Paleozoic pteridosperm cuticles it has shown increased contrast, emphasized cutinization differences, and enhanced the three-dimensional morphology of complex epidermal features when compared with other dyes. The compound has also been used with alcian blue in the histochemical study of the distribution and morphometric characteristics of mast cells. It has also been used to detect mucin and cartilage.

Solubility
Soluble in water at 20°C, 50g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 477-73-6
Molecular Formula C20H19ClN4
Molecular Weight (g/mol) 350.85
MDL Number MFCD00011759
InChI Key QRYAEWIQIBAZOJ-UHFFFAOYSA-N
Synonym basic red 2, safranine o, gossypimine, safranin, safranine t, safranin o, safranin t, safranine, tolusafranine, hidaco safranine
PubChem CID 2723800
IUPAC Name 3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride
SMILES [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1

Specifications

Odor Odorless
Quantity 50 g
Beilstein 3924099
Solubility Information Soluble in water at 20°C,50g/L. Soluble in alcohol.
Formula Weight 350.85
Chemical Name or Material Safranine O
Hazard Category H315-H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
EINECSNumber 207-518-8
RTECSNumber SG1623000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only