missing translation for 'onlineSavingsMsg'
Learn More

Safflower oil, Carthamus tinctorius

Catalog No. AAJ63982K2
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63982K2 1 L
AAJ63982AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63982K2 Supplier Thermo Scientific Chemicals Supplier No. J63982K2

CAS: 8001-23-8

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Solubility
Insoluble in cold water and hot water.

Notes
Supplied as a raw oil without preservative

Chemical Identifiers

CAS 8001-23-8
MDL Number MFCD00132216
Synonym Carthamus tinctorius oil

Specifications

Color Yellow
Density 0.921
Flash Point >113°C (235°F)
Refractive Index 1.476
Quantity 1 L
Solubility Information Insoluble in cold water and hot water.
Physical Form Oil
Chemical Name or Material Safflower oil, Carthamus tinctorius
EINECSNumber 232-276-5
RTECSNumber VN2230000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only