Learn More
Sol-Care™ Safety Lancets
Catalog No. 361060397
Change view
Click to view available options
Needle Gauge:
21 G
23 G
28 G
30 G
Length (Metric):
1.6 mm
1.8 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (Metric)
361060397 21 G 1.8 mm
361060343 30 G 1.6 mm
361060333 28 G 1.6 mm
361060392 28 G 1.8 mm
361060349 23 G 1.8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 361060397 Supplier Sol-Care™ Supplier No. SLA008
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Safety Lancet is a sterile, single use, springe-loaded lancet designed for capillary blood sampling. The lancet is precision sharpened designed for maximum comfort and optimal blood flow.
  • The needle is safely concealed in the lancet body before and after use
  • Easy twist-off cap removal
  • Comfortable finger recess design
  • Small contact face for precise puncturing
Specifications
Disposable Single-use
Material Polyethylene
Needle Gauge 21 G
For Use With (Equipment) Blood Collection Tube Holder
Length (Metric) 1.8 mm
Sterility Non-sterile
Product Type Lancet