missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Safety-Labeled Wash Bottles
GREENER_CHOICE

Catalog No. FB0340923G
Change view
Click to view available options
Chemical Label:
Deionized Water
Distilled Water
Unlabeled
Water
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Label
FB0340923G Water
FB0340923M Deionized Water
FB0340923N Unlabeled
FB0340923K Distilled Water
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FB0340923G Supplier Fisherbrand™ Supplier No. FB0340923G
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Fisherbrand™ Safety-Labeled Wash Bottles also include Chemical Abstract Service (CAS) number, protective equipment symbols and target organ inf.

  • Easy-to-read printed bottles
  • Wide mouth 53mm (2 in.) bottle for easy refilling
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Polypropylene closures
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200

Use only with solvents compatible with LDPE.

Specifications

Capacity (Metric) 500 mL
Color Natural
Material Polyethylene
Closure Color Blue
Mouth Wide
Chemical Label Water
Closure Material Polypropylene
Closure Size 53 mm
Autoclavable Not Autoclavable
Capacity (English) 17 oz.
Certifications/Compliance OSHA 29 CFR 1910.1200
Show More Show Less