missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Safety Blood Collection Set

Catalog No. 22040224
Change view
Click to view available options
Size:
21 G
23 G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22040224 23 G
22040223 21 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22040224 Supplier Greiner Bio-One Supplier No. 450185

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sterile and preassembled for ease of use

  • Cost effective; ensures your patient only has to undergo one venipuncture for all procedures
  • Allows collection of blood into blood culture bottles and Vacuette™ blood collection tubes in one procedure
  • Safety Blood Collection Set with preattached holder has been designed for use with most commonly used blood culture bottles
  • Compatible with BacT/Alert and BacTec blood culture bottles
  • Standard Blood Collection Set (without safety machanism) is also available with preassembled blood culture holder; contact your Fisher HealthCare Representative
Note: After blood collection, promptly dispose of in an approved sharps disposal container in accordance with the procedures of your facility.

Specifications

Size 23 G
Length (Metric) Tube 30 cm
Length (English) Tube 11.81 in.
For Use With (Equipment) Blood culture bottles
Certifications/Compliance CE, ISO 11014
Color Clear
Latex Free Yes
Product Type Safety Blood Collection Set
Sterility Sterile