missing translation for 'onlineSavingsMsg'
Learn More

S-(Thiobenzoyl)thioglycolic Acid 97.0+%, TCI America™

Catalog No. T24015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24015G 5 g
T240125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24015G Supplier TCI America Supplier No. T24015G
Only null left

Chemical Identifiers

CAS 942-91-6
Molecular Formula C9H8O2S2
Molecular Weight (g/mol) 212.28
MDL Number MFCD00004927
InChI Key XBEIANFIOZTEDE-UHFFFAOYSA-N
Synonym (Thiobenzoylthio)acetic Acid
PubChem CID 70336
IUPAC Name 2-(benzenecarbothioylsulfanyl)acetic acid
SMILES OC(=O)CSC(=S)C1=CC=CC=C1

Specifications

Melting Point 128°C
Quantity 5 g
Formula Weight 212.28
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material S-(Thiobenzoyl)thioglycolic Acid