missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide 98.0+%, TCI America™

Catalog No. A16991G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16991G Supplier TCI America Supplier No. A16991G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 167316-27-0
Molecular Formula C21H23N2O2S
Molecular Weight (g/mol) 367.49
MDL Number MFCD03095684
InChI Key UOPFIWYXBIHPIP-SFTDATJTSA-O
Synonym 1s,2s-+-n-p-tosyl-1,2-diphenylethylenediamine, s,s-tsdpen, 1s,2s-+-n-4-toluenesulfonyl-1,2-diphenylethylenediamine, 1s,2s-n-4-toluenesulfonyl-1,2-diphenylethylene-1,2-diamine, 1s,2s---n-p-tosyl-1,2-diphenylethylenediamine, n-1s,2s-2-amino-1,2-diphenylethyl-4-methylbenzenesulfonamide, 1s, 2s-+-n-4-toluene sulfonyl 1,2-diphenyl-1,2-ethane diamine, s,s-n-p-toluenesulfonyl-1,2-diphenylethylenediamine, 1s,2s-tsdpen, s,s-n-2-amino-1,2-diphenylethyl-p-toluenesulfonamide
PubChem CID 6612782
IUPAC Name N-[(1S,2S)-2-azaniumyl-1,2-diphenylethyl]-4-methylbenzene-1-sulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N[C@H]([C@@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 1 g
Formula Weight 366.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide