missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-(+)-2,3-Bis(tert-butylmethylphosphino)quinoxaline 98.0+%, TCI America™

Catalog No. B5302100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5302100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5302100MG Supplier TCI America Supplier No. B5302100MG

Chemical Identifiers

CAS 1107608-80-9
Molecular Formula C18H28N2P2
Molecular Weight (g/mol) 334.384
MDL Number MFCD10567042
InChI Key DRZBLHZZDMCPGX-UHFFFAOYSA-N
Synonym (S,S)-QuinoxP*
PubChem CID 16218188
IUPAC Name tert-butyl-[3-[tert-butyl(methyl)phosphanyl]quinoxalin-2-yl]-methylphosphane
SMILES CC(C)(C)P(C)C1=NC2=CC=CC=C2N=C1P(C)C(C)(C)C

Specifications

Melting Point 102°C
Quantity 100 mg
Formula Weight 334.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-(+)-2,3-Bis(tert-butylmethylphosphino)quinoxaline