missing translation for 'onlineSavingsMsg'
Learn More

(S)-(p-Toluenesulfinyl)ferrocene 97.0+%, TCI America™

Catalog No. T25461G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T25461G Supplier TCI America Supplier No. T25461G

Chemical Identifiers

CAS 164297-25-0
Molecular Formula C17H16FeOS
Molecular Weight (g/mol) 324.219
InChI Key QAAXXGKGRWQHLR-FMOMHUKBSA-N
Synonym (S)-Ferrocenyl p-Tolyl Sulfoxide
PubChem CID 15472425
IUPAC Name cyclopenta-1,3-diene;1-[(R)-cyclopenta-1,3-dien-1-ylsulfinyl]-4-methylbenzene;iron(2+)
SMILES CC1=CC=C(C=C1)S(=O)C2=CC=C[CH-]2.[CH-]1C=CC=C1.[Fe+2]

Specifications

Melting Point 143°C
Quantity 1 g
Formula Weight 324.22
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(p-Toluenesulfinyl)ferrocene