missing translation for 'onlineSavingsMsg'
Learn More

(S)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine 97.0+%, TCI America™

Catalog No. D2536100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2536100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2536100MG Supplier TCI America Supplier No. D2536100MG

Chemical Identifiers

CAS 55650-58-3
Molecular Formula C26H28FeNP
Molecular Weight (g/mol) 441.34
MDL Number MFCD00075285
InChI Key RCAFPSZHKFOLEE-KVEBOLLKNA-N
Synonym s-+-n,n-dimethyl-1-2-diphenylphosphino ferrocenylethylamine, s-+-n,n-dimethyl-1-2-diphenylphosphino ferrocenyl ethylamine, s-+-n,n-dimethyl-1-r-2-diphenylphosphino ferrocenyl ethylamine, s-ppfa, +-s-n,n-dimethyl-1-r-2-diphenylphosphino ferrocenyl ethylamine
IUPAC Name (S)-(+)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine
SMILES [Fe].c1cccc1.C[C@H](N(C)C)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 145°C
Quantity 100 mg
Formula Weight 441.34
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine