missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98%, 98% ee, Thermo Scientific Chemicals

Catalog No. AC441095000
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC441095000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC441095000 Supplier Thermo Scientific Chemicals Supplier No. 441095000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1172127-44-4 | Molecular Formula: C25H22FNO4 | Molecular Weight: 419.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1172127-44-4
Molecular Formula C25H22FNO4
Molecular Weight (g/mol) 419.45
MDL Number MFCD17019320
InChI Key VNYGHKCAXQKZEQ-VWLOTQADSA-N
Synonym s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-fluorophenyl-2-methylpropanoic acid, s-n-fmoc-alpha-methyl-2-fluorophenylalaine, fmoc-alpha-methyl-d-2-fluorophe, fmoc-alpha-methyl-d-2-fluorophenylalanine, r-n-fmoc-a-methyl-2-fluorophenylalanine
PubChem CID 44598473
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(2-fluorophenyl)-2-methylpropanoic acid
SMILES CC(CC1=C(F)C=CC=C1)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 122.0°C to 125.0°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Specific Rotation − 47.00
Formula Weight 419.46
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (S)-N-FMOC-α-Methyl-2-fluorophenylalanine, 98% ee
Show More Show Less

RUO – Research Use Only